MilliporeSigma
  • The ins and outs of calreticulin: from the ER lumen to the extracellular space.
The ins and outs of calreticulin: from the ER lumen to the extracellular space.
Trends in cell biology (2001-04-18)
S Johnson, M Michalak, M Opas, P Eggleton
ABSTRACT
Calreticulin was first isolated 26 years ago. Since its discovery as a minor Ca(2+)-binding protein of the sarcoplasmic reticulum, the appreciation of its importance has grown, and it is now recognized to be a multifunctional protein, most abundant in the endoplasmic reticulum (ER). The protein has well-recognized physiological roles in the ER as a molecular chaperone and Ca(2+)-signalling molecule. However, it has also been found in other membrane-bound organelles, at the cell surface and in the extracellular environment, where it has recently been shown to exert a number of physiological and pathological effects. Here, we will focus on these less-well-characterized functions of calreticulin.