MilliporeSigma
[Efficacy of fenticonazole in patients with pityriasis versicolor].
Giornale italiano di dermatologia e venereologia : organo ufficiale, Societa italiana di dermatologia e sifilografia (1989-09-01)
A Di Silverio, M Mosca, G Brandozzi, G P Vignoli, M Gatti, P Gabba, S Ubezio
ABSTRACT
Thirty patients suffering from Pityriasis versicolor were treated with fenticonazole in cream or lotion form with two applications a day. At microscopy, Malassezia furfur was encountered in 25 cases and Pityrosporum orbiculare in 5. Disappearance of the yeast was obtained on average in 2 weeks of treatment. Measurement of sebum content with a Sebumeter apparatus did not reveal significant difference (p greater than 0.05) between patients suffering from Pityriasis versicolor and controls and in treated patients, in the course of topical therapy.
Product Description
Fenticonazole nitrate, European Pharmacopoeia (EP) Reference Standard