MilliporeSigma
  • The sesquiterpenes polygodial and drimanial in vitro affect glutamatergic transport in rat brain.
The sesquiterpenes polygodial and drimanial in vitro affect glutamatergic transport in rat brain.
Neurochemical research (2006-05-31)
Lucia Helena Martini, Lucas Cereser, Isaac Zanonato Junior, Fluvia Melina Alves Jardim, Deusa Aparecida Vendite, Marcos Emilio dos Santos Frizzo, Rosendo A Yunes, João Batista Calixto, Susana Wofchuk, Diogo O Souza
ABSTRACT
Natural products including those derived from plants, have over the years greatly contributed to the development of therapeutic drugs. Polygodial and drimanial are sesquiterpenes isolated from the bark of the plant Drymis Winteri (Winteraceae) that exhibit antinociceptive properties. Since peripheral glutamate presents nociceptive actions, in this study it was investigated the effects of hydroalcooholic extracts from Drymis winteri (polygodial and drimanial) on the glutamatergic system in rat brain. Polygodial and drimanial inhibited glutamate uptake by astrocytes, as well as by cortical, hippocampal and striatal slices, and increased synaptosomal glutamate release. These concurrent effects would predispose to an increase in the extracellular glutamate concentrations, leading to possible neurotoxic effects (excitotoxicity) of these natural compounds, which would suggest the need for some caution in their therapeutic application.
Sigma-Aldrich
Polygodial, ≥97% (HPLC)