MilliporeSigma
  • Rates of cortisol increase and decrease in channel catfish and sunshine bass exposed to an acute confinement stressor.
Rates of cortisol increase and decrease in channel catfish and sunshine bass exposed to an acute confinement stressor.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2006-02-10)
Kenneth B Davis, Brian C Small
ABSTRACT
Channel catfish and sunshine bass were exposed to a low-water stress event and allowed to recover in fresh water or a solution of metomidate (dl-1-(1-phenylethyl)-5-(metoxycarbonyl) imidazole hydrochloride), which inhibits the synthesis of cortisol. Change in time of plasma cortisol was used as an index of cortisol secretion and clearance. Plasma cortisol and glucose increased during the exposure to low-water stress in both fish, but the changes of both plasma components were more dramatic in sunshine bass. Exposure to metomidate during recovery resulted in a short-term increase in plasma glucose but differences between controls and metomidate-exposed fish were relatively minor thereafter. Cortisol began to decrease in catfish immediately after the removal of the stress but continued to increase for 15 min in sunshine bass recovering in fresh water and for 5 min in bass recovering in metomidate. Catfish recovering in fresh water had a cortisol elimination rate of -1.28 ng/mL/min compared with -2.45 ng/mL/min for fish recovering in metomidate (P>0.05) while sunshine bass recovering in fresh water had an elimination rate of -6.96 ng/mL/min compared with -4.50 ng/mL/min for fish recovering in metomidate (P>0.05). These data indicate that the rapid decrease of plasma cortisol after removal of the stressor is due to an almost immediate decrease of secretion, tissue uptake and a rapid renal loss due to the absence of a plasma binding protein.
Product Description
Etomidate impurity B, European Pharmacopoeia (EP) Reference Standard