MilliporeSigma
  • Structure of the DNA repair and replication endonuclease and exonuclease FEN-1: coupling DNA and PCNA binding to FEN-1 activity.
Structure of the DNA repair and replication endonuclease and exonuclease FEN-1: coupling DNA and PCNA binding to FEN-1 activity.
Cell (1998-10-20)
D J Hosfield, C D Mol, B Shen, J A Tainer
ABSTRACT
Flap endonuclease (FEN-1) removes 5' overhanging flaps in DNA repair and processes the 5' ends of Okazaki fragments in lagging strand DNA synthesis. The crystal structure of Pyrococcus furiosus FEN-1, active-site metal ions, and mutational information indicate interactions for the single- and double-stranded portions of the flap DNA substrate and identify an unusual DNA-binding motif. The enzyme's active-site structure suggests that DNA binding induces FEN-1 to clamp onto the cleavage junction to form the productive complex. The conserved FEN-1 C terminus binds proliferating cell nuclear antigen (PCNA) and positions FEN-1 to act primarily as an exonuclease in DNA replication, in contrast to its endonuclease activity in DNA repair. FEN-1 mutations altering PCNA binding should reduce activity during replication, likely causing DNA repeat expansions as seen in some cancers and genetic diseases.