MilliporeSigma
Grhl3 and Lmo4 play coordinate roles in epidermal migration.
Developmental biology (2008-07-16)
Nikki R Hislop, Jacinta Caddy, Stephen B Ting, Alana Auden, Sumitha Vasudevan, Sarah L King, Geoffrey J Lindeman, Jane E Visvader, John M Cunningham, Stephen M Jane
ABSTRACT
In addition to its role in formation of the epidermal barrier, the mammalian transcription factor Grainy head-like 3 (Grhl3) is also essential for neural tube closure and wound repair, processes that are dependent in part on epidermal migration. Here, we demonstrate that the LIM-only domain protein, LMO4 serves as a functional partner of GRHL3 in its established roles, and define a new cooperative role for these factors in another developmental epidermal migration event, eyelid fusion. GRHL3 and LMO4 interact biochemically and genetically, with mutant mice exhibiting fully penetrant exencephaly, thoraco-lumbo-sacral spina bifida, defective skin barrier formation, and a co-incident eyes-open-at-birth (EOB) phenotype, which is not observed in the original individual null lines. The two genes are co-expressed in the surface ectoderm of the migrating eyelid root, and electron microscopy of Grhl3/Lmo4-null eyes reveals a failure in epithelial extension and a lack of peridermal clump formation at the eyelid margins. Accumulation of actin fibers is also absent in the circumference of these eyelids, and ERK1/2 phosphorylation is lost in the epidermis and eyelids of Grhl3(-/-)/Lmo4(-/-) embryos. Keratinocytes from mutant mice fail to "heal" in in vitro scratch assays, consistent with a general epidermal migratory defect that is dependent on ERK activation and actin cable formation.