MilliporeSigma
  • Amplification of the ch19p13.2 NACC1 locus in ovarian high-grade serous carcinoma.
Amplification of the ch19p13.2 NACC1 locus in ovarian high-grade serous carcinoma.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2011-01-18)
Ie-Ming Shih, Kentaro Nakayama, Gang Wu, Naomi Nakayama, Jinghui Zhang, Tian-Li Wang
ABSTRACT
On the basis of digital karyotyping, we have identified a new, discrete amplified region at ch19p13.2 in a high-grade ovarian serous carcinoma. To further characterize this region, we determined the frequency and biological significance of ch19p13.2 amplification by analyzing 341 high-grade serous carcinomas from The Cancer Genome Atlas (TCGA) and found an increased DNA copy number at this locus in 18% of cases. We correlated the DNA and RNA copy number by analyzing the TCGA data set for all amplified genes and detected seven genes within ch19p13.2 that were significantly correlated (R≥0.54) and were, in fact, listed as the top 100 potential 'driver' genes at a genome-wide scale. Interestingly, one of the seven genes, NACC1, encoding NAC1 was previously reported to be involved in the development of tumor recurrence in ovarian serous carcinoma and to have a causal role in the development of paclitaxel resistance. Therefore, we selected NACC1 for validation in an independent cohort. On the basis of fluorescence in situ hybridization, we found that 35 (20%) of 175 high-grade serous carcinomas had an increased DNA copy number at the NACC1 locus, and those amplified cases were associated with early disease recurrence within 6 months (P=0.013). A significantly high level of NAC1 protein expression based on immunohistochemistry was detected in amplified tumors as compared with non-amplified tumors (P<0.005). In summary, our data suggest that amplification at the ch19p13.2 NACC1 locus, leading to NAC1 overexpression, is one of the molecular genetic alterations associated with early tumor recurrence in ovarian cancer.