MilliporeSigma
Antibody-dependent enhancement of Marburg virus infection.
The Journal of infectious diseases (2011-10-19)
Eri Nakayama, Daisuke Tomabechi, Keita Matsuno, Noriko Kishida, Reiko Yoshida, Heinz Feldmann, Ayato Takada
ABSTRACT
Marburg virus (MARV) and Ebola virus (EBOV) cause severe hemorrhagic fever in primates. Earlier studies demonstrated that antibodies to particular epitopes on the glycoprotein (GP) of EBOV enhanced virus infectivity in vitro. To investigate this antibody-dependent enhancement (ADE) in MARV infection, we produced mouse antisera and monoclonal antibodies (mAbs) to the GPs of MARV strains Angola and Musoke. The infectivity of vesicular stomatitis virus pseudotyped with Angola GP in K562 cells was significantly enhanced in the presence of Angola GP antisera, whereas only minimal ADE activity was seen with Musoke GP antisera. This difference correlated with the percentage of hybridoma clones producing infectivity-enhancing mAbs. Using mAbs to MARV GP, we identified 3 distinct ADE epitopes in the mucinlike region on Angola GP. Interestingly, some of these antibodies bound to both Angola and Musoke GPs but showed significantly higher ADE activity for strain Angola. ADE activity depended on epitopes in the mucinlike region and glycine at amino acid position 547, present in the Angola but absent in the Musoke GP. These results suggest a possible link between ADE and MARV pathogenicity and provide new insights into the mechanisms underlying ADE entry of filoviruses.
Product Description
Custom Peptides, Product Overview, Design, and Ordering