MilliporeSigma
  • IGHV3-21 gene usage is associated with high TCL1 expression in chronic lymphocytic leukemia.
IGHV3-21 gene usage is associated with high TCL1 expression in chronic lymphocytic leukemia.
European journal of haematology (2009-11-06)
Mahmoud R Mansouri, Marie Sevov, Anna Aleskog, Mikael Jondal, Mats Merup, Christer Sundström, Lyda Osorio, Richard Rosenquist
ABSTRACT
T-cell leukemia/lymphoma protein 1 (TCL1) was recently shown to display an expression pattern in chronic lymphocytic leukemia (CLL) corresponding to molecular subtypes, where poor-risk patients demonstrated higher expression levels. Here, we examined the mRNA expression pattern of TCL1 in 144 patients with CLL, including 67 immunoglobulin heavy-chain variable (IGHV) mutated, 58 IGHV unmutated and 19 patients with IGHV3-21 usage. A higher TCL1 expression level was detected in patients with CLL with unmutated vs. mutated IGHV genes (P < 0.001), whereas no difference was demonstrated within the IGHV3-21 cohort (i.e., mutated vs. unmutated and stereotyped vs. non-stereotyped complementarity determining region 3). The IGHV3-21 subgroup displayed high TCL1 mRNA expression, differing significantly from other IGHV mutated cases (P < 0.001), although 11/19 had mutated IGHV genes. Furthermore, high TCL1 expression levels were associated with significantly shorter overall survival (P < 0.001). Altogether, we show that TCL1 mRNA expression may predict clinical outcome in CLL and that the IGHV3-21 subset, regardless of mutational status, displays high TCL1 expression.