MilliporeSigma
  • Interleukin-6 and the serotonergic system of the medulla oblongata in the sudden infant death syndrome.
Interleukin-6 and the serotonergic system of the medulla oblongata in the sudden infant death syndrome.
Acta neuropathologica (2009-04-28)
Ingvar Jon Rognum, Robin L Haynes, Ashild Vege, May Yang, Torleiv O Rognum, Hannah C Kinney
ABSTRACT
Mild infection may trigger sudden death in the vulnerable infant by cytokine interactions with a compromised medullary serotonergic (5-HT) system, leading to disrupted cardiorespiratory regulation and sleep-related sudden death. The cytokine interleukin (IL)-6 is elevated in the cerebrospinal fluid in SIDS. We tested the hypothesis that the expression of IL-6 receptors (IL-6R) and/or gp130 (involved in IL-6R signaling) is altered in the medullary 5-HT system in SIDS. Immunohistochemistry of IL-6R and gp130 was performed on medullae from 25 SIDS infants, 20 infectious deaths, and 14 controls using a semi-quantitative grading system. In the SIDS cases, mean IL-6R intensity grade in the arcuate nucleus (major component of medullary 5-HT system) was significantly higher than in the control group (2.00 +/- 0.07 vs. 1.77 +/- 0.08, P = 0.04), with no other differences in IL-6R or gp130 expression at any other site. Arcuate 5-HT neurons expressed IL-6R, indicating a site of IL-6/5-HT interaction. In SIDS, IL-6R expression is abnormal in the arcuate nucleus, the putative human homolog of rodent ventral medullary chemosensitivity sites involving 5-HT. Aberrant interactions between IL-6 and the arcuate nucleus may contribute to impaired responses to hypercapnia generated by infection (hyper-metabolism) combined with rebreathing.