MilliporeSigma
Caveolin-1 in breast cancer.
Cancer biology & therapy (2004-11-13)
Toula Bouras, Michael P Lisanti, Richard G Pestell
ABSTRACT
Caveolin-1 is the principal structural protein of caveolae, sphingolipid and cholesterol-rich invaginations of the plasma membrane involved in vesicular trafficking and signal transduction. During caveolae-dependent signaling, caveolin-1 acts as a scaffold protein to sequester and organize multi-molecular signaling complexes involved in diverse cellular activities and, as such serves as a paradigm by which numerous disease processes may be affected by ablation or mutation of caveolin-1. The hypothesis that caveolin-1 conveys a tumor/transformation suppressor function in the mammary gland is derived from several independent lines of evidence accumulated by genetic, molecular and clinical approaches. The human caveolin-1 gene maps to a suspected tumor suppressor locus (D7S522/7q31.1) frequently deleted in human breast carcinomas. In addition, up to 16% of human breast carcinomas harbor a dominant-negative mutation, P132L, in the caveolin-1 gene. Caveolin-1 RNA and protein levels are also downregulated in human primary breast carcinomas and cell lines, with reintroduction of caveolin-1 in vitro sufficient to inhibit numerous tumorigenic properties, including anchorage independent growth and invasiveness. Most recently caveolin-1 knockout mice have provided breakthroughs in understanding the dynamic role of caveolin-1 in the pathogenesis of mammary epithelial cell hyperplasia, tumorigenesis and metastasis in a vivo setting. This review concentrates on recent advances implicating caveolin-1 in breast cancer pathogenesis, with emphasis on the signaling pathways regulated during these processes.