MilliporeSigma
Fungal biotransformation of 6-nitrochrysene.
Applied and environmental microbiology (1998-08-04)
J V Pothuluri, J B Sutherland, J P Freeman, C E Cerniglia
ABSTRACT
The fungus Cunninghamella elegans was used to biotransform 6-nitrochrysene, a mutagen that is a widespread environmental contaminant. After 6 days, 74% of the 3H-labeled 6-nitrochrysene added had been metabolized to two isomeric sulfate conjugates. These conjugates were separated by high-performance liquid chromatography and identified by UV-visible, 1H nuclear magnetic resonance, and mass spectral techniques as 6-nitrochrysene 1-sulfate and 6-nitrochrysene 2-sulfate.
Product Description
6-Nitrochrysene, BCR®, certified reference material