MilliporeSigma
  • A review of clinical studies of bropirimine immunotherapy of carcinoma in situ of the bladder and upper urinary tract.
A review of clinical studies of bropirimine immunotherapy of carcinoma in situ of the bladder and upper urinary tract.
European urology (1997-01-01)
M F Sarosdy
ABSTRACT
Bropirimine has been shown to have activity against carcinoma in situ (CIS) of the bladder in a previous phase-I trial. A review of three completed clinical trials as well as ongoing studies is presented to provide a current update. Details of the initial phase-I trial are reviewed, as are findings of a subsequent phase-II trial in bladder CIS and a multicenter study in upper tract CIS. All have used a dose of 3 g/day for 3 consecutive days each week, repeated weekly for up to 1 year. Cytology must be positive prior to treatment, and both biopsies and cytology must be negative after therapy for the patient to be considered a complete response. In the phase-II trial in bladder CIS, 20 (61%) of 33 patients had a complete response. Responders included patients with prior bacillus Calmette-Guérin (BCG) therapy, uni- and multifocal CIS, and primary and secondary CIS. Responses were seen in 10 (48%) of 21 evaluable patients with upper tract CIS. Toxicities in both studies were manageable in most patients. Trials underway include bropirimine in BCG-failed CIS, a randomized comparison to BCG in previously untreated patients, and a trial of the two together. Bropirimine does have activity against both bladder and upper tract CIS on the dose schedule used to date.
Sigma-Aldrich
Bropirimine, ≥98% (HPLC)