MilliporeSigma
  • miR-211 promotes the progression of head and neck carcinomas by targeting TGFβRII.
miR-211 promotes the progression of head and neck carcinomas by targeting TGFβRII.
Cancer letters (2013-06-04)
Ting-Hui Chu, Cheng-Chieh Yang, Chung-Ji Liu, Mann-Tin Lui, Shu-Chun Lin, Kuo-Wei Chang
ABSTRACT
miR-211 up-regulation and transforming growth factor-β type II receptor (TGFβRII) down-regulation are associated with poor prognosis of head and neck squamous cell carcinoma (HNSCC). miR-211 directly targets TGFβRII with the miR-211-TGFβRII-c-Myc axis promoting HNSCC progression. An inverse correlation of miR-211 and TGFβRII expression was found in metastatic HNSCC samples. After 4-nitroquinoline 1-oxide induction, more severe epithelial tumorigenesis was detected on K14-miR-211 transgenic mouse dorsal tongues. Human metastatic lesions and mouse tongue tumors showed increased nuclear c-Myc expression. A novel role for miR-211 in the regulation of TGFβRII and c-Myc during tumorigenesis being revealed should help to develop anti-HNSCC therapies.
Sigma-Aldrich
4-Nitroquinoline N-oxide, ≥98%