MilliporeSigma
  • In vitro and in vivo antitumor effects of the essential oil from the leaves of Guatteria friesiana.
In vitro and in vivo antitumor effects of the essential oil from the leaves of Guatteria friesiana.
Planta medica (2012-01-26)
Anny C S Britto, Allan C A de Oliveira, Raissa M Henriques, Gabriella M B Cardoso, Diogo S Bomfim, Adriana A Carvalho, Manoel O Moraes, Claudia Pessoa, Maria L B Pinheiro, Emmanoel V Costa, Daniel P Bezerra
ABSTRACT
Guatteria friesiana (W. A. Rodrigues) Erkens & Maas (synonym Guatteriopsis friesiana W. A. Rodrigues), popularly known as "envireira", is a medicinal plant found in the Brazilian and Colombian Amazon basin that is used in traditional medicine for various purposes. Recent studies on this species have demonstrated antimicrobial activity. In this study, the antitumor activity of the essential oil from the leaves of G. friesiana (EOGF) and its main components ( α-, β-, and γ-eudesmol) were determined using experimental models. In the in vitro study, EOGF and its components α-, β-, and γ-eudesmol displayed cytotoxicity against tumor cell lines, showing IC₅₀ values in the range of 1.7 to 9.4 µg/mL in the HCT-8 and HL-60 cell lines for EOGF, 5.7 to 19.4 µg/mL in the HL-60 and MDA-MB-435 cell lines for α-eudesmol, 24.1 to > 25 µg/mL in the SF-295 and MDA-MB-435 cell lines for β-eudesmol, and 7.1 to 20.6 µg/mL in the SF-295 and MDA-MB-435 cell lines for γ-eudesmol, respectively. In the in vivo study, the antitumor effect of EOGF was evaluated in mice inoculated with sarcoma 180 tumor cells. Tumor growth inhibition rates were 43.4-54.2 % and 6.6-42.8 % for the EOGF treatment by intraperitoneal (50 and 100 mg/kg/day) and oral (100 and 200 mg/kg/day) administration, respectively. The treatment with EOGF did not significantly affect body mass, macroscopy of the organs, or blood leukocyte counts. Based on these results, we can conclude that EOGF possesses significant antitumor activity and has only low systemic toxicity. These effects could be assigned to its components α-, β-, and γ-eudesmol.
Sigma-Aldrich
β-Eudesmol, ≥90% (GC)