MilliporeSigma
  • Extraction of cellulose-synthesizing activity of Gluconacetobacter xylinus by alkylmaltoside.
Extraction of cellulose-synthesizing activity of Gluconacetobacter xylinus by alkylmaltoside.
Carbohydrate research (2011-11-11)
Akira Hashimoto, Kenji Shimono, Yoshiki Horikawa, Tsukasa Ichikawa, Masahisa Wada, Tomoya Imai, Junji Sugiyama
ABSTRACT
This study reinvestigated the synthesis of cellulose in vitro with a well-known cellulose-producing bacterium, Gluconacetobacter xylinus. Alkylmaltoside detergents, which are more frequently used in recent structural biological researches, are uniquely used in this study to solubilize cellulose-synthesizing activity from the cell membrane of G. xylinus. Activity comparable to that previously reported is obtained, while the synthesized cellulose is crystallized into a non-native polymorph of cellulose (cellulose II) as well as the previous studies. In spite of this failure to recover the native activity to synthesize cellulose I microfibril in vitro, the product is a polymer with a degree of polymerization greater than 45 as determined by matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOFMS). It was thus concluded that the established protocol can solubilize cellulose-synthesizing activity of G. xylinus with polymerizing activity.
Sigma-Aldrich
Digitonin, ~50% (TLC)
Sigma-Aldrich
Digitonin, Used as non-ionic detergent