MilliporeSigma
  • Determination of memantine in rat plasma by HPLC-fluorescence method and its application to study of the pharmacokinetic interaction between memantine and methazolamide.
Determination of memantine in rat plasma by HPLC-fluorescence method and its application to study of the pharmacokinetic interaction between memantine and methazolamide.
Biomedical chromatography : BMC (2011-05-20)
Mohamed G Hassan, Kamla M Emara, Horria A Mohamed, Hanaa M Abdel-Wadood, Rie Ikeda, Mitsuhiro Wada, Naotaka Kuroda, Kenichiro Nakashima
ABSTRACT
A sensitive high-performance liquid chromatographic method with fluorescence detection was developed to determine memantine (MT) in rat plasma. The method consists of pre-column labeling of MT with 4-(4,5-diphenyl-1H-imidazol-2-yl)benzoyl chloride (DIB-Cl) and a clean-up step with solid-phase extraction. A good separation of DIB-MT was achieved within 12 min on an octadecylsilica (ODS) column (150 × 4.6 mm i.d.; 5 µm) with a mobile phase of acetonitrile-water (70:30, v/v). The calibration curve prepared with fluoxetine as an internal standard showed good linearity in the range of 10-400 ng/mL (r = .999). The limits of detection and quantitation at signal-to-noise ratios of 3 and 10 were 2.0 and 6.6 ng/mL, respectively. The method was shown to be reliable with precisions of <5% for intra-day and <9% for inter-day as relative standard deviation. The fluorescence property and reaction yield of authentic DIB-MT were also examined. The proposed method was successfully applied to study the pharmacokinetic interaction between MT and methazolamide.