MilliporeSigma
  • One year intermittent rifaximin plus fibre supplementation vs. fibre supplementation alone to prevent diverticulitis recurrence: a proof-of-concept study.
One year intermittent rifaximin plus fibre supplementation vs. fibre supplementation alone to prevent diverticulitis recurrence: a proof-of-concept study.
Digestive and liver disease : official journal of the Italian Society of Gastroenterology and the Italian Association for the Study of the Liver (2012-10-25)
Angel Lanas, Julio Ponce, Angelo Bignamini, Fermín Mearin
ABSTRACT
Evidence supporting appropriate medical therapy to prevent recurrence of colonic diverticulitis is limited. Our goal was to evaluate the potential of rifaximin given periodically in addition to fibre for the prophylaxis of recurrences. We conducted a multicentre, randomized, open controlled study in patients with a recent episode of colonic diverticulitis, currently in remission. Patients received 3.5 g of high-fibre supplementation b.d. with or without one week per month of the non-absorbable antibiotic rifaximin (400 mg b.d.) for 12months. Primary endpoint was recurrence of diverticulitis, encompassing acute symptomatic flare with or without complications, analyzed by multivariable logistic regression analysis and by Cox proportional hazard method. After randomizing 165 patients, the study was interrupted since the recruitment rate was largely below the minimum anticipated, and the trial was switched from evidence-gathering to proof-of-concept. Recurrences occurred in 10.4% of patients given rifaximin plus fibres vs. 19.3% of patients receiving fibres alone. The logistic analysis adjusted for sex, age, illness duration, time from last episode, disease localization and centre recruitment rate, yielded a significant treatment effect (odds ratio 3.20; 95% confidence interval: 1.16-8.82; P=0.025). Patients with diverticulitis diagnosed since ≥1 year receiving rifaximin also had a lower incidence of recurrences (10%; 95% confidence interval: 2-47% vs. 67%; 95% confidence interval: 37-100%). Both treatments were safe. This study represents a proof-of concept of the efficacy of cyclic rifaximin treatment, added to fibre supplements, to reduce the risk of recurrences of diverticulitis in patients in remission.
Product Description
Supelco
Rifaximin, VETRANAL®, analytical standard
Sigma-Aldrich
Rifaximin