MilliporeSigma
  • Distribution and sources of organochlorinated contaminants in sediments from Izmir Bay (Eastern Aegean Sea).
Distribution and sources of organochlorinated contaminants in sediments from Izmir Bay (Eastern Aegean Sea).
Marine pollution bulletin (2011-04-06)
I Pazi, F Kucuksezgin, L T Gonul
ABSTRACT
Eighteen surface sediment samples representative of the entire Izmir Bay were analyzed for organochlorine pesticides (OCPs) and polychlorinated biphenyls (PCBs). This was in order to provide extensive information concerning the recent deposition of these compounds in this area, together with levels, distribution, possible sources and potential biological risk. Sites that were contaminated by high concentrations of organochlorine compounds were associated with dense population, such as the harbour and the Gediz River estuary. Relatively higher ΣDDT concentrations and high DDT/DDE+DDD ratios in the Gediz River estuary indicated DDT usage, probably linked to public health emergencies. According to the established guidelines for sediment quality, the risk of adverse biological effects from such levels of OCPs and PCBs as recorded at most of the study sites was insignificant. But the higher concentrations in Inner bay and in the Gediz River estuary could cause biological damage.