MilliporeSigma
  • Preparation of zwitterionic hybrid polymer and its application for the removal of heavy metal ions from water.
Preparation of zwitterionic hybrid polymer and its application for the removal of heavy metal ions from water.
Journal of hazardous materials (2010-03-13)
Junsheng Liu, Yue Ma, Tongwen Xu, Guoquan Shao
ABSTRACT
A series of zwitterionic hybrid polymers were prepared from the ring-opening polymerization of pyromellitic acid dianhydride (PMDA) and phenylaminomethyl trimethoxysilane (PAMTMS), and a subsequent sol-gel process. FTIR spectra confirmed the step products. TGA analysis showed that the thermal degradation temperature increased with an increase in PMDA content. As a typical example, sample B was used to separate Cu(2+) and Pb(2+) removal by adsorption. It was indicated that its adsorption for Cu(2+) and Pb(2+) followed Lagergren second-order kinetic model and Langmuir isotherm model, demonstrating that the adsorption process might be Langmuir monolayer adsorption. Meanwhile, it was found that the adsorption capacity of Pb(2+) on sample B is beyond 12 times higher than that of Cu(2+) in 0.1 mol dm(-3) aqueous solution, revealing that it has larger affinity for Pb(2+). The desorption efficiency of Cu(2+) and Pb(2+) in 1 mol dm(-3) HNO(3) solution reached up to 96 and 89%, respectively; indicating that they can be regenerated and recycled in industry. These findings suggest that they are promising adsorbents for the selective removal of Pb(2+) from Pb(2+)/Cu(2+) mixed aqueous solution, and can be applied to separate and recover the heavy metal ions from contaminated water and waste chemicals.
Sigma-Aldrich
Pyromellitic dianhydride, 97%