MilliporeSigma
  • A semisynthetic glycosphingolipid (LIGA20) reduces 2,4, 5-trihydroxyphenylalanine neurotoxicity in primary neuronal cultures.
A semisynthetic glycosphingolipid (LIGA20) reduces 2,4, 5-trihydroxyphenylalanine neurotoxicity in primary neuronal cultures.
European journal of pharmacology (1993-10-12)
S D Skaper, E Fadda, L Facci, H Manev
ABSTRACT
The semisynthetic glycosphingolipid derivative II3Neu5-AcGgOse4-2-d-erythro-1,3-dihydroxy-2-chloro-acetamid e-4-trans- octadacene (LIGA20) attenuated injury induced by the excitotoxic L-dopa metabolite 2,4,5-trihydroxyphenylalanine (TOPA) in cultures of rat cerebellar granule cells when presented simultaneously with TOPA (EC50; 9 microM LIGA20). The natural glycosphingolipid ganglioside GM1 up to 200 microM was not neuroprotective as cotreatment, although pretreatment of cells for 2 h was efficacious. This greater potency and speed of LIGA20 action extended to limiting TOPA-induced death of cultured mesencephalic dopaminergic neurons. These data suggest that LIGA20 may have therapeutic potential for the treatment of disorders associated with excitotoxic processes.
Product Description
USP
Levadopa Related Compound A, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
6-Hydroxy-DL-DOPA, ≥98% (HPLC), powder