MilliporeSigma
  • Purification, properties and cation activation of galactosyltransferase from lactating-rat mammary Golgi membranes.
Purification, properties and cation activation of galactosyltransferase from lactating-rat mammary Golgi membranes.
European journal of biochemistry (1988-02-01)
N Navaratnam, S Ward, C Fisher, N J Kuhn, J N Keen, J B Findlay
ABSTRACT
Galactosyltransferase was purified from Golgi membranes of lactating-rat mammary gland and studied with respect to its physical and enzymic (lactose synthetase) properties. The enzyme occurred in both monomeric (43-46 kDa) and apparently dimeric (90 kDa) forms. It was very unstable except in the presence of phospholipid, detergent, or cations binding to site 2. The amino acid composition and the N-terminal sequence closely resembled that of the human and bovine milk enzymes, particularly in respect to a Pro-Pro-Pro-Pro sequence. Kinetic studies demonstrated a high-affinity Mn2+-binding site (1) essential for activity, and a low-affinity Mn2+-binding site (2) that could also bind spermidine or clupeine. Mn2+ binding at site 2 raised Vmax fivefold. Spermidine binding at site 2 enhanced Mn2+ binding at site 1, and influenced binding of glucose. At physiological glucose concentration, clupeine or spermidine activated nearly as well as 15 mM MnCl2 and are regarded as models of a natural cation activator that remains to be isolated. Evidence is given for an essential histidine residue in the galactosyltransferase. It is proposed that site 1 Mn2+ participates directly in the reaction mechanism, whereas site 2 is a regulator site allosterically activated by a basic protein.
Sigma-Aldrich
Protamine sulfate salt from herring, Grade III, histone, free(Millon test)