MilliporeSigma
  • Interspecific variations of cerebral endothelial cholinesterases in rodents and carnivores.
Interspecific variations of cerebral endothelial cholinesterases in rodents and carnivores.
Brain research (1989-01-09)
J Trancard, I S Delamanche, G Ruiz, C Bouchaud
ABSTRACT
The cholinesterase equipment of cerebral microvessels was studied in some rodents and carnivores using the Koelle-Friedenwald histochemical method with 3 artificial substrates and specific inhibitors for butyrylcholinesterase or acetylcholinesterase. Our observations reveal a great heterogeneity in cholinesterase types and their distribution in each of the different species studied. Only in the rat, butyrylcholinesterase appears to be a marker for the microvessels provided with a blood-brain barrier.
Sigma-Aldrich
Propionylthiocholine iodide, ≥98%