MilliporeSigma
  • Efficient ex vivo induction of T cells with potent anti-tumor activity by protein antigen encapsulated in nanoparticles.
Efficient ex vivo induction of T cells with potent anti-tumor activity by protein antigen encapsulated in nanoparticles.
Cancer immunology, immunotherapy : CII (2013-04-25)
Rodney A Rosalia, Ana Luisa Silva, Marcel Camps, Ahmed Allam, Wim Jiskoot, Sjoerd H van der Burg, Ferry Ossendorp, Jaap Oostendorp
ABSTRACT
Protein antigen (Ag)-based immunotherapies have the advantage to induce T cells with a potentially broad repertoire of specificities. However, soluble protein Ag is generally poorly cross-presented in MHC class I molecules and not efficient in inducing robust cytotoxic CD8(+) T cell responses. In the present study, we have applied poly(lactic-co-glycolic acid) (PLGA) nanoparticles (NP) which strongly improve protein Ag presentation by dendritic cells (DC) in the absence of additional Toll-like receptor ligands or targeting devices. Protein Ag-loaded DC were used as antigen presenting cells to stimulate T cells in vitro and subsequently analyzed in vivo for their anti-tumor effect via adoptive transfer, a treatment strategy widely studied in clinical trials as a therapy against various malignancies. In a direct comparison with soluble protein Ag, we show that DC presentation of protein encapsulated in plain PLGA-NP results in efficient activation of CD4(+) and CD8(+) T cells as reflected by high numbers of activated CD69(+) and CD25(+), interferon (IFN)-γ and interleukin (IL)-2-producing T cells. Adoptive transfer of PLGA-NP-activated CD8(+) T cells in tumor-bearing mice displayed good in vivo expansion capacity, potent Ag-specific cytotoxicity and IFN-γ cytokine production, resulting in curing mice with established tumors. We conclude that delivery of protein Ag through encapsulation in plain PLGA-NP is a very efficient and simple procedure to stimulate potent anti-tumor T cells.