MilliporeSigma
  • In vitro cytocompatibility evaluation of alginate dialdehyde for biological tissue fixation.
In vitro cytocompatibility evaluation of alginate dialdehyde for biological tissue fixation.
Carbohydrate polymers (2012-12-12)
Yuanting Xu, Li Li, Hao Wang, Xixun Yu, Zhipeng Gu, Chengcheng Huang, Hong Peng
ABSTRACT
Biological tissues must be chemically fixed before they can be implanted in humans, due to the immediate degradation and presence of antigenicity of naturally derived tissues. To provide a crosslinking reagent which is cytocompatible and may prepare biocompatible fixed tissues, a novel crosslinking agent, alginate dialdehyde (ADA), was employed to fix biological tissues by our group. The study was to evaluate the cytocompatibility of ADA for biological tissue fixation. Glutaraldehyde and genipin counterparts were used as controls. The result suggested that the cytotoxicity of ADA was significantly lower than that of glutaraldehyde and genipin. Additionally, in the evaluation of cytotoxicity of fixed tissue itself and the residues, as well as the cell adhesion property, ADA-fixed tissue was significantly superior to its glutaraldehyde counterpart and comparable to its genipin counterpart. The results obtained in this study demonstrate that ADA is a cytocompatible crosslinking reagent for biological tissue fixation.
Sigma-Aldrich
Genipin, ≥98% (HPLC), powder