MilliporeSigma
  • Reproductive and developmental toxicity screening test of 3-cyanopyridine in rats.
Reproductive and developmental toxicity screening test of 3-cyanopyridine in rats.
Reproductive toxicology (Elmsford, N.Y.) (2012-11-21)
Mika Takahashi, Kaoru Yabe, Hina Kato, Tomoko Kawamura, Mariko Matsumoto, Mutsuko Hirata-Koizumi, Atsushi Ono, Akihiko Hirose
ABSTRACT
Crl:CD(SD)rats were given 3-cyanopyridine by gavage at 0, 5, 30 or 180 mg/kg/day. Males were dosed for 42 days beginning 14 days before mating, and females for 40-53 days beginning 14 days before mating to day 3 of lactation, including throughout the mating and gestation periods. General toxicity, mainly liver damage, was observed in males at ≥30 mg/kg/day and in females at ≥5 mg/kg/day. Sertoli cell vacuolation was observed at 180 mg/kg/day, and spermatocyte damages were observed at ≥30 mg/kg/day. Effects on estrous cycles, corpora lutea and implantations, and unsuccessfully mated females, despite additional mating, were observed at 180 mg/kg/day. Delayed initiation of delivery, dystocia, and deaths or moribundities of pregnant females were observed at 180 mg/kg/day, and only two pregnant rats delivered live pups at that dose. The NOAEL for reproductive/developmental toxicity was concluded to be 30 mg/kg/day.
Sigma-Aldrich
3-Pyridinecarbonitrile, 98%