MilliporeSigma
  • Pinacolborane as the boron source in nitrogen-directed borylations of aromatic N,N-dimethylhydrazones.
Pinacolborane as the boron source in nitrogen-directed borylations of aromatic N,N-dimethylhydrazones.
The Journal of organic chemistry (2012-10-18)
Rocío López-Rodríguez, Abel Ros, Rosario Fernández, José M Lassaletta
ABSTRACT
A mild procedure for the Ir(III)-catalyzed nitrogen-directed ortho borylation of aromatic N,N-dialkylhydrazones using pinacolborane as the boron source has been developed. The methodology relies on a modified, hemilabile N,N ligand built on a 4-N,N-dimethylaminopyridine unit that provides high reactivity while maintaining exclusive ortho-selectivity. This procedure can be combined with Suzuki-Miyaura cross-couplings in a 'one-pot' fashion to afford functionalized biaryl derivatives that, upon subsequent 'one-pot', high yielding transformations, provide a convenient entry for the preparation of advanced benzonitrile intermediates for the synthesis of Sartan-type drugs.
Product Description
Supelco
Benzonitrile, suitable for HPLC, 99.9%
Sigma-Aldrich
Benzonitrile, ReagentPlus®, 99%