MilliporeSigma
  • Effect of timolol on refractive outcomes in eyes with myopic regression after laser in situ keratomileusis: a prospective randomized clinical trial.
Effect of timolol on refractive outcomes in eyes with myopic regression after laser in situ keratomileusis: a prospective randomized clinical trial.
American journal of ophthalmology (2012-09-01)
Ahmad Shojaei, Medi Eslani, Yasaman Vali, Mohammad Mansouri, Nima Dadman, Mehdi Yaseri
ABSTRACT
To compare the effects of timolol on refractive outcomes in eyes with myopic regression after laser in situ keratomileusis (LASIK) with a control-matched group. Prospective, randomized, parallel-controlled, double-masked clinical trial. A computer-generated randomization list based on random block permutation (length 4 to 8) was used for treatment allocation. setting: Basir Eye Center, Tehran, Iran. Of 124 eyes with myopic regression after LASIK using Technolas 217-Z, 45 eyes in each group were analyzed. Patients were randomly assigned into either Group 1, who received timolol 0.5% eye drops, or Group 2, who received artificial tears for 6 months. Spherical equivalent (SE) at 6 months posttreatment. In Group 1, SE improved from -1.48 ± 0.99 diopter (D) before treatment to -0.88 ± 0.91 D and -0.86 ± 0.93 D 6 months after treatment and 6 months after timolol discontinuation, respectively (P < .001). In Group 2, it was -1.57 ± 0.67 D, -1.83 ± 0.76 D, and -1.91 ± 0.70 D, respectively (P < .001). SE was significantly better in Group 1 6 months after treatment and 6 months after discontinuation of treatment (P < .001 for both comparisons). There was a 0.26 D decrease in SE improvement every 4 months after the surgery in the Group 1 (P < .001). Timolol application is effective for the treatment of myopic regression after LASIK compared with control group. Its effects last for at least 6 months after its discontinuation.
Sigma-Aldrich
Timolol maleate salt, ≥98% (TLC), powder