MilliporeSigma
  • Protective effect of deferricoprogen isolated from Monascus purpureus NTU 568 on citrinin-induced apoptosis in HEK-293 cells.
Protective effect of deferricoprogen isolated from Monascus purpureus NTU 568 on citrinin-induced apoptosis in HEK-293 cells.
Journal of agricultural and food chemistry (2012-07-28)
Li-Chuan Hsu, Ya-Wen Hsu, Yu-Han Liang, Zhi-Hu Lin, Yao-Haur Kuo, Tzu-Ming Pan
ABSTRACT
Monascus species have traditionally been used in Asian food, with rice as their fermentation substrate. Red mold rice (RMR) contains citrinin, a nephrotoxic agent capable of exerting oxidative stress and cellular apoptosis. We investigated the components in RMR that could minimize the adverse effects of citrinin. Combining chemical separations and bioactivity assays, we identified an antioxidative component called deferricoprogen (DFC) in the fermented rice of Monascus purpureus NTU 568. The DFC structure was confirmed by nuclear magnetic resonance (NMR) and mass spectra analysis. The 2,2-diphenyl-1-picrylhydrazyl (DPPH) free-radical-scavenging activity of DFC was similar to that of vitamin E. The 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl-tetrazolium bromide (MTT) assay and flow cytometric analysis showed the effect of DFC and citrinin on cell viability and cell cycle. DFC was found to be protective against the cytotoxicity and cell death induced by citrinin on human embryonic kidney (HEK-293) cells. DFC also demonstrated anti-apoptotic property in preventing citrinin-induced apoptosis.
Product Description
Supelco
Citrinin solution, ~100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Citrinin, from Penicillium citrinum, ≥98% (HPLC)