MilliporeSigma
  • Neutral zone approach for the rehabilitation of a severely atrophic mandibular ridge: Case report.
Neutral zone approach for the rehabilitation of a severely atrophic mandibular ridge: Case report.
General dentistry (2012-05-25)
Komal Ladha, Shubhra Gill, Rekha Gupta, Mahesh Verma
ABSTRACT
Providing a complete denture for a patient with an atrophic mandibular ridge presents a unique challenge for dentists. Retention, stability, and support must be considered when fabricating and fitting a complete denture. This article highlights the use of the neutral zone technique for successful complete denture therapy in a patient with an atrophic mandibular ridge, with special emphasis on the role of coverage of genial tubercles for maximum prosthesis support and stability.
Sigma-Aldrich
Poly(ethyl methacrylate), average Mw ~515,000 by GPC, powder