MilliporeSigma
  • Inflammation-induced effects on iron-related proteins in splenic macrophages and the liver in mice.
Inflammation-induced effects on iron-related proteins in splenic macrophages and the liver in mice.
Blood cells, molecules & diseases (2012-04-17)
Abitha Sukumaran, Aparna Venkatraman, Molly Jacob
ABSTRACT
Anemia of inflammation is characterized by disturbances in systemic iron homeostasis. In order to better understand the events involved, we carried out a time-course study on the effects of acute and chronic inflammation on iron-related proteins in mouse splenic macrophages and the liver. Mice were sacrificed at various time points ranging from 0 h up to 4 weeks after induction of inflammation with turpentine oil. Expression levels of iron-related proteins in the splenic macrophages and liver were determined. Iron levels in the serum, spleen and liver were also measured. Hepatic hepcidin was found to be induced in response to inflammation. In the macrophages, expression levels of ferroportin and TfR1 were decreased at some of the time points. The expression of hepatic TfR1 and ferritin was significantly higher at the early time points. Ferritin levels in the liver decreased progressively thereafter; this was associated with significantly higher ferroportin expression in the liver, despite high levels of hepcidin, suggesting that hepcidin may not regulate ferroportin levels in the liver, unlike in the macrophages. The effects of hepcidin, thus, appeared to be tissue-specific. Serum iron levels were decreased initially; these then rose and were associated with decreasing iron levels in the liver and spleen. Thus, inflammation affected the expression levels of many proteins involved in iron homeostasis in splenic macrophages and the liver, with differences seen in the effects at these 2 sites. These effects are likely to contribute to the development of anemia of inflammation.
Sigma-Aldrich
Oil of turpentine, purified