MilliporeSigma
  • An investigation of the correlation between blood concentration of mizoribine and its efficacy in treatment of rheumatoid arthritis based on indices of drug survival and improvement in DAS28-CRP.
An investigation of the correlation between blood concentration of mizoribine and its efficacy in treatment of rheumatoid arthritis based on indices of drug survival and improvement in DAS28-CRP.
Modern rheumatology (2012-03-07)
Hideo Ohtsubo, Sanae Ohkura, Masaki Akimoto, Yoshiya Arishima, Nobuhiko Sunahara, Tomomaro Izumihara, Nobutaka Eiraku, Tamami Yoshitama, Takemasa Matsuda
ABSTRACT
The efficacy of mizoribine (MZR) in treatment of rheumatoid arthritis (RA) was retrospectively investigated in terms of drug survival, improvement in Disease Activity Score-28 (DAS28)-C-reactive protein (CRP), and blood MZR concentration obtained 3 h after dosing (MZR-C3). To compare the efficacy of MZR administered via different regimens, the subjects were divided into 2 groups: those receiving a single dose of MZR at 100-150 mg every other day (group A) and those receiving 2 or 3 divided doses of the drug on consecutive days, which is the usual dosing method of the drug (group B). Group A had significantly higher MZR-C3 levels compared with group B, as well as significantly greater improvement in DAS28-CRP. Moreover, drug survival was significantly longer in group A. The primary regression equation suggested that the effective blood MZR concentration in RA treatment is MZR-C3 of 1.47 µg/mL or more. The results of the present study indicate that it is possible to increase the efficacy of MZR in a blood concentration-dependent manner, and also to control RA over a prolonged period, using single administration of MZR on alternate days at an increased dose.
Sigma-Aldrich
Mizoribine, ≥98% (TLC)