MilliporeSigma
  • Iron-catalysed tandem cross-dehydrogenative coupling (CDC) of terminal allylic C(sp3) to C(sp2) of styrene and benzoannulation in the synthesis of polysubstituted naphthalenes.
Iron-catalysed tandem cross-dehydrogenative coupling (CDC) of terminal allylic C(sp3) to C(sp2) of styrene and benzoannulation in the synthesis of polysubstituted naphthalenes.
Chemical communications (Cambridge, England) (2012-02-07)
Hua Liu, Li Cao, Jia Sun, John S Fossey, Wei-Ping Deng
ABSTRACT
A novel iron-catalysed tandem cross-dehydrogenative coupling and benzoannulation process was developed for the synthesis of biologically and synthetically important polysubstituted naphthalene derivatives from simple 1,2-aryl-propenes and styrenes in moderate to good yields.