MilliporeSigma
  • The cholecystokinin CCK2 receptor antagonist, JNJ-26070109, inhibits gastric acid secretion and prevents omeprazole-induced acid rebound in the rat.
The cholecystokinin CCK2 receptor antagonist, JNJ-26070109, inhibits gastric acid secretion and prevents omeprazole-induced acid rebound in the rat.
British journal of pharmacology (2012-02-04)
T D Barrett, G Lagaud, P Wagaman, J M Freedman, W Yan, L Andries, M C Rizzolio, M F Morton, N P Shankley
ABSTRACT
JNJ-26070109 [(R)4-bromo-N-[1-(2,4-difluoro-phenyl)-ethyl]-2-(quinoxaline-5-sulfonylamino)-benzamide] is a novel antagonist at cholecystokinin CCK(2) receptors with good pharmacokinetic properties and represents a novel mechanism for the treatment of gastro-oesophageal reflux disease (GORD). The purpose of the present study was to determine whether chronic treatment with JNJ-26070109 could prevent, as well as treat, acid rebound in rats. A chronic fistula was surgically inserted into the stomach of rats to enable the measurement of acid secretion under basal, pentagastrin and histamine-stimulated conditions. JNJ-26070109 and omeprazole were administered separately and in combination. Sustained administration of omeprazole alone and in combination with JNJ-26070109 inhibited gastric acid secretion by >90%. However, 3 days after withdrawing treatment, there was a rebound hypersecretion by ∼1.5-fold in omeprazole-treated animals. No such acid rebound was observed with JNJ-26070109 alone or with co-administration of JNJ-26070109 and omeprazole. The anti-trophic effects of JNJ-26070109 in the gastric mucosal paralleled the effects on acid rebound. Administration of JNJ-26070109 for 3 days after cessation of omeprazole prevented the occurrence of acid rebound. Interestingly, chronic, but not acute, treatment with JNJ-26070109 also inhibited histamine-stimulated acid secretion. Chronic administration of JNJ-26070109 effectively inhibited gastric acid secretion and suppressed proton pump inhibitor (PPI)-induced acid rebound in the rat. This work advances the field by demonstrating that modest doses of a competitive CCK(2) receptor antagonist have significant and functionally important anti-trophic actions in the gastric mucosa. These properties make JNJ-26070109 a suitable candidate for clinical investigation for the treatment of GORD.
Sigma-Aldrich
Pentagastrin, ≥95% (HPLC), powder