MilliporeSigma
  • [Effect of polycyclic aromatic hydrocarbons on laccase production by white rot fungus Pleurotus ostreatus D1].
[Effect of polycyclic aromatic hydrocarbons on laccase production by white rot fungus Pleurotus ostreatus D1].
Prikladnaia biokhimiia i mikrobiologiia (2012-01-12)
N N Pozdniakova, S V Nikiforova, O E Makarov, O V Turkovskaia
ABSTRACT
The effect of polycyclic aromatic hydrocarbons (PAHs) on the dynamics of laccase production by the fungus Pleurotus ostreatus D1 under conditions of submerged cultivation on Kirk's medium has been studied. It has been shown that phenanthrene, fluoranthene, pyrene, and chrysene actively induce this enzyme, whereas fluorene and anthrecene had a smaller effect. Addition of Mn2+ ions to cultivation medium elevates the laccase activity twofold and more in the presence of all the studied PAHs. Electrophoresis under nondenaturing conditions demonstrates induction of additional laccase species by xenobiotics. Ligninolytic peroxidase activities are undetectable under the conditions used.
Product Description
Supelco
Chrysene, analytical standard
Chrysene, BCR®, certified reference material
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland