MilliporeSigma
A contemporary update on 'soft' linings.
Dental update (2011-04-20)
J Fraser McCord, Alan C Donaldson, Thomas James Lamont
ABSTRACT
This article presents a contemporary overview of soft or compliant denture lining materials. It does not focus on denture bases, which are intrinsically flexible, and suggests some clinical options which readers might wish to consider. The aim of this article is to update clinicians on the types of compliant linings currently available and to advise on how some might be used.
Sigma-Aldrich
Poly(ethyl methacrylate), average Mw ~515,000 by GPC, powder