MilliporeSigma
  • Schiglautone A, a new tricyclic triterpenoid with a unique 6/7/9-fused skeleton from the stems of Schisandra glaucescens.
Schiglautone A, a new tricyclic triterpenoid with a unique 6/7/9-fused skeleton from the stems of Schisandra glaucescens.
Organic letters (2011-02-23)
Fan-Yu Meng, Jia-Xiang Sun, Xue Li, Heng-Yi Yu, Shu-Ming Li, Han-Li Ruan
ABSTRACT
Schiglautone A (1), a unique 6/7/9-fused tricyclic carbon backbone triterpenoid, was isolated from the stems of Schisandra glaucescens. Its structure was determined on the basis of spectroscopic analysis and single-crystal X-ray crystallography. A hypothetical biosynthetic pathway of 1 was postulated.
Sigma-Aldrich
Schizandrin, ≥98% (HPLC)