MilliporeSigma
  • Kinetic determination of tellurium based on its inhibitory effect on the palladium(II)-catalysed reaction between pyronine G and hypophosphite ion.
Kinetic determination of tellurium based on its inhibitory effect on the palladium(II)-catalysed reaction between pyronine G and hypophosphite ion.
The Analyst (1990-09-01)
C Sánchez-Pedreño, M I Albero, M Soledad García, A Saez
ABSTRACT
A kinetic method for the determination of Te based on its inhibitory effect on the PdII-catalysed reaction between pyronine G and H2PO2- is described. The influence of experimental variables on the rate of the process and the potential interfering effect of a large number of ions has been studied. Under the selected experimental conditions: 6 x 10(-5) M pyronine G; 0.6 M H2PO2-; pH 2.6, adjusted with Britton-Robinson buffer; 0.80 microgram ml-1 of PdII; and a temperature of 22 +/- 0.2 degrees C, Te was determined in the concentration range 0.08-0.85 microgram ml-1. The method was applied to the determination of Te in waters and lead concentrates.
Sigma-Aldrich
Sodium hypophosphite, puriss., meets analytical specification of BPC63, anhydrous, 98-101%
Sigma-Aldrich
Sodium hypophosphite monohydrate, ≥99%