MilliporeSigma
  • Antileishmanial and antitrypanosomal activities of the 8-aminoquinoline tafenoquine.
Antileishmanial and antitrypanosomal activities of the 8-aminoquinoline tafenoquine.
Antimicrobial agents and chemotherapy (2010-09-15)
Vanessa Yardley, Francisco Gamarro, Simon L Croft
ABSTRACT
The 8-aminoquinoline tafenoquine showed significant in vitro activity against Leishmania species, including L. donovani amastigotes in macrophages, with 50% inhibitory concentrations (IC(50)s) between 0.1 and 4.0 μM for both pentavalent antimony (SbV)-sensitive and SbV-resistant strains and by oral administration in BALB/c mice, with 50% effective dose (ED(50)) values of 1.2 to 3.5 mg/kg for 5 days. Tafenoquine was less active against intracellular Trypanosoma cruzi amastigotes, with an IC(50) of 21.9 μM.
Sigma-Aldrich
Tafenoquine succinate, ≥95% (HPLC)