MilliporeSigma
  • Estimation of ambient GABA levels in layer I of the mouse neonatal cortex in brain slices.
Estimation of ambient GABA levels in layer I of the mouse neonatal cortex in brain slices.
The Journal of physiology (2010-04-28)
Anton Dvorzhak, Olga Myakhar, Petr Unichenko, Knut Kirmse, Sergei Kirischuk
ABSTRACT
GABAergic synapses on Cajal-Retzius neurons in layer I of the murine neocortex experience GABA(B) receptor (GABA(B)R)-mediated tonic inhibition. Extracellular GABA concentration ([GABA](o)) that determines the strength of GABA(B)R-mediated inhibition is controlled by GABA transporters (GATs). In this study, we hypothesized that the strength of presynaptic GABA(B)R activation reflects [GABA](o) in the vicinity of synaptic contacts. Slices obtained from two age groups were used, namely postnatal days (P)2-3 and P5-7. GABAergic postsynaptic currents (IPSCs) were recorded using the whole-cell patch-clamp technique. Minimal electrical stimulation in layer I was applied to elicit evoked IPSCs (eIPSCs) using a paired-pulse protocol. Three parameters were selected for comparison: the mean eIPSC amplitude, paired-pulse ratio, and failure rate. When GAT-1 and GAT-2/3 were blocked by NO-711 (10 microM) and SNAP-5114 (40 microM), respectively, no tonic GABA(B)R-mediated inhibition was observed. In order to restore the control levels of GABA(B)R-mediated inhibition, 250 and 125 nm exogenous GABA was required at P2-3 and P5-7, respectively. Addition of 3-mercaptopropionic acid, a glutamate decarboxylase inhibitor, did not significantly change the obtained values arguing against the suggestion that a mechanism different from GATs contributes to [GABA](o) control. We conclude that juxtasynaptic [GABA](o) is higher (about 250 nM) at P2-3 than at P5-7 (about 125 nM). As both radial cell migration and corticogenesis in general are strongly dependent on [GABA](o) and the formation of the last layer 2/3 is finished by P4 in rodents, the observed [GABA](o) reduction in layer I might reflect this crucial event in the cortical development.
Sigma-Aldrich
NO-711 hydrochloride, ≥98% (HPLC)