MilliporeSigma
  • Silent increase of urinary ethylmalonic acid is an indicator of nonspecific brain dysfunction.
Silent increase of urinary ethylmalonic acid is an indicator of nonspecific brain dysfunction.
NMR in biomedicine (2010-02-27)
Raffaella Zannolli, Sabrina Buoni, Maria Tassini, Anna De Nicola, Gianni Betti, Claudio De Felice, Alessandra Orsi, Maria Concetta Varetti, Francesco Ferrara, Mario Messina, Cosimo Giannini, Angelika Mohn, Francesco Chiarelli, Marco Liberati, Mirella Strambi, Silvia Funghini, Antonio Vivi, Ron A Wevers, Joseph Hayek
ABSTRACT
Our aim was to compare urinary ethylmalonic acid (EMA) levels in subjects who had no apparent clinical reason to have increased levels of this substance but were suffering from non-specific CNS impairment, and healthy controls. Urinary EMA concentrations detected by (1)H-NMR spectroscopy were studied in 130 subjects with CNS impairment of unknown origin (with no definite diagnosis, no specific symptoms or signs, and normal common biochemical and metabolic screening results) and 130 age- and sex-matched healthy subjects. EMA levels exceeding two standard deviations (SD) above normal (i.e. 8.1 mmol/molCn) were found in a subgroup of CNS-impaired patients and healthy controls. EMA levels exceeding 2 SD above normal were fourfold prevalent in the urine of patients with non-specific CNS impairment compared to from the EMA levels in healthy controls. Moreover, we found that the level exceeding > 8.1 mmol/molCn (i.e. > + 2 SD) had sufficient discrimination accuracy in identifying subjects with non-specific CNS impairment; the level exceeding 12 mmol/molCn (i.e. > + 6 SD) reaches suitable accuracy (i.e. 100% specificity and 78.6% sensitivity). These observations are of importance, as we found that subtle increases in urinary EMA levels are frequent in patients with non-specific CNS impairment. The reasons for this association remain unknown.
Sigma-Aldrich
Ethylmalonic acid, 97%