MilliporeSigma
[Late onset 3-HMG-CoA lyase deficiency: a rare but treatable disorder].
Archives de pediatrie : organe officiel de la Societe francaise de pediatrie (2009-11-26)
S Pierron, H Giudicelli, M Moreigne, A Khalfi, G Touati, C Caruba, M-O Rolland, C Acquaviva
ABSTRACT
3-Hydroxy-3-methylglutaric aciduria is a rare autosomal recessive genetic disorder due to a deficiency of the 3-hydroxy-3-methylglutarylCoA lyase (HMG-CoA lyase), a mitochondrial enzyme involved in ketogenesis and in the final step of l-leucine catabolism. HMG-CoA lyase deficiency can lead, in particular circumstances, such as fever, prolonged fasting or digestive disorders, to brutal and severe hypoglycemia with metabolic acidosis and sometimes fatal coma. We report on a new case of 3-hydroxy-3-methylglutaric aciduria particular by its late onset in a 3-year-old patient. Molecular investigation identified two new sequence modifications in the HMGCL gene: c.494G>A (p.Arg165Gln) and c.820G>A (p.Gly274Arg). We remind about this case report that the therapeutical is mainly preventive and allows a very good prognosis for this disease. Long-term treatment consists in limited fasting time, continuous low protein diet and l-carnitine supplementation. Preventive measures are essential: prevention of fasting and emergency treatment during intercurrent infections.
Sigma-Aldrich
3-Hydroxy-3-methylglutaric acid, ≥95%