MilliporeSigma
AU-rich elements regulate Drosophila gene expression.
Molecular and cellular biology (2009-03-11)
Fatima Cairrao, Anason S Halees, Khalid S A Khabar, Dominique Morello, Nathalie Vanzo
ABSTRACT
In mammals, AU-rich elements (AREs) are critical regulators of mRNA turnover. They recruit ARE-binding proteins that inhibit or stimulate rapid mRNA degradation in response to stress or developmental cues. Using a bioinformatics approach, we have identified AREs in Drosophila melanogaster 3' untranslated regions and validated their cross-species conservation in distant Drosophila genomes. We have generated a Drosophila ARE database (D-ARED) and established that about 16% of D. melanogaster genes contain the mammalian ARE signature, an AUUUA pentamer in an A/U-rich context. Using candidate ARE genes, we show that Drosophila AREs stimulate reporter mRNA decay in cultured cells and in the physiological context of the immune response in D. melanogaster. In addition, we found that the conserved ARE-binding protein Tis11 regulates temporal gene expression through ARE-mediated decay (AMD) in D. melanogaster. Our work reveals that AREs are conserved and functional cis regulators of mRNA decay in Drosophila and highlights this organism as a novel model system to unravel in vivo the contribution of AMD to various processes.
Sigma-Aldrich
Cecropin A, ≥97% (HPLC), powder