MilliporeSigma
  • Novel three-component route to diastereoselective synthesis of trisubstituted vinylphosphonates using phosphites, acetylenic esters, and aroyl chlorides.
Novel three-component route to diastereoselective synthesis of trisubstituted vinylphosphonates using phosphites, acetylenic esters, and aroyl chlorides.
Journal of combinatorial chemistry (2008-12-23)
Sadegh Rostamnia, Abdolali Alizadeh, Long-Guan Zhu
ABSTRACT
Trisubstituted vinylphosphonates have been prepared via three-component reaction using phosphites, acetylenic esters, and aroyl chlorides in good yields. A variety of phosphites, activated acetylenes, and aroyl chlorides have been successfully employed in these reactions. In addition, three-component synthesis of vinylphosphonate provides exclusive E-olefin stereochemistry.
Sigma-Aldrich
Vinylphosphonic acid, ≥90% (T)
Sigma-Aldrich
Vinylphosphonic acid, 97%