MilliporeSigma
  • [Effects of solution pH and simulated acid rain on the behavior of two sulfonylurea herbicides in soil].
[Effects of solution pH and simulated acid rain on the behavior of two sulfonylurea herbicides in soil].
Ying yong sheng tai xue bao = The journal of applied ecology (2007-06-08)
Wei Zhang, Jin-Jun Wang
ABSTRACT
By the methods of batch equilibration and leaching, this paper studied the effects of solution pH and simulated acid rain on the behavior of bensulfuron-methyl and metsulfuron-methyl in soil. The results showed that the adsorption isotherms of these two herbicides fitted Freundlich equation well, and their adsorbed amounts reduced obviously with the increasing pH of water-soil system, which in turn promoted the translocation of the herbicides in soil. The adsorption coefficient (Kf) was positively correlated with soil organic matter and clay contents, while negatively correlated with soil pH. The higher pH of simulated acid rain had a greater contribution on the leaching of the two sulfonylurea herbicides, and their leached amount was increased with increasing acid rain. There was a close relationship between the leaching of the herbicides and the properties of soil. The soils with higher contents of organic matter and clay had a greater retention capability to the herbicides.
Product Description
Supelco
Bensulfuron-methyl, PESTANAL®, analytical standard