MilliporeSigma
  • Practical and convenient synthesis of coumarins from phenols and propiolic acid esters.
Practical and convenient synthesis of coumarins from phenols and propiolic acid esters.
Nature protocols (2007-04-21)
Tsugio Kitamura, Juzo Oyamada, Toshiyuki Tsubota
ABSTRACT
This protocol describes the synthesis of 6,7-methylenedioxy-4-phenylcoumarin from sesamol and ethyl phenylpropiolate using a Pd(OAc)2 catalyst to illustrate coumarin synthesis. This procedure is simple and easy and can be applied to the synthesis of other coumarins that have electron-rich phenol groups. The reaction is conducted by stirring a solution of Pd(OAc)2, sesamol and ethyl phenylpropiolate in trifluoroacetic acid at room temperature (15-20 degrees C) under atmospheric conditions. This protocol can be completed in 3 d.
Sigma-Aldrich
Ethyl phenylpropiolate, 98%