MilliporeSigma
  • Chemoenzymatic synthesis of trans-dihydrodiol derivatives of monosubstituted benzenes from the corresponding cis-dihydrodiol isomers.
Chemoenzymatic synthesis of trans-dihydrodiol derivatives of monosubstituted benzenes from the corresponding cis-dihydrodiol isomers.
Organic & biomolecular chemistry (2007-01-26)
Derek R Boyd, Narain D Sharma, Nuria M Llamas, Gerard P Coen, Peter K M McGeehin, Christopher C R Allen
ABSTRACT
Enantiopure trans-dihydrodiols have been obtained by a chemoenzymatic synthesis from the corresponding cis-dihydrodiol metabolites, obtained by dioxygenase-catalysed arene cis-dihydroxylation at the 2,3-bond of monosubstituted benzene substrates. This generally applicable, seven-step synthetic route to trans-dihydrodiols involves a regioselective hydrogenation and a Mitsunobu inversion of configuration at C-2, followed by benzylic bromination and dehydrobromination steps. The method has also been extended to the synthesis of both enantiomers of the trans-dihydrodiol derivatives of toluene, through substitution of a vinyl bromine atom of the corresponding trans-dihydrodiol enantiomers derived from bromobenzene. Through incorporation of hydrogenolysis and diMTPA ester diastereoisomer resolution steps into the synthetic route, both trans-dihydrodiol enantiomers of monohalobenzenes were obtained from the cis-dihydrodiols of 4-haloiodobenzenes.
Sigma-Aldrich
Vinyl bromide solution, 1.0 M in THF