MilliporeSigma
  • Bafoudiosbulbins A, and B, two anti-salmonellal clerodane diterpenoids from Dioscorea bulbifera L. var sativa.
Bafoudiosbulbins A, and B, two anti-salmonellal clerodane diterpenoids from Dioscorea bulbifera L. var sativa.
Phytochemistry (2006-08-01)
Rémy Bertrand Teponno, Azefack Léon Tapondjou, Donatien Gatsing, Jules Désiré Djoukeng, Eliane Abou-Mansour, Raphael Tabacchi, Pierre Tane, H Stoekli-Evans, David Lontsi
ABSTRACT
Two clerodane diterpenoids, Bafoudiosbulbins A 1, and B 2, together with five known compounds: tetracosanoic acid, 1-(tetracosanoyl)-glycerol, trans-tetracosanylferulate, beta-sitosterol and 3-O-beta-D-glucopyranosyl-beta-sitosterol were isolated from the tubers of Dioscorea bulbifera L. var sativa. Their structures were established by spectroscopic methods (1D and 2D-NMR, MS) and X-ray crystallographic diffraction analysis of compound 1. The CH2Cl2-soluble portion of the crude extract and the two clerodanes were screened for anti-bacterial activity using both agar diffusion and broth dilution techniques against Pseudomonas aeruginosa, Escherichia coli, Klebsiella pneumoniae, Staphylococcus aureus, Salmonella typhi, Salmonella paratyphi A and Salmonella paratyphi B. They both showed significant activities against P. aeruginosa, S. typhi, S. paratyphi A and S. paratyphi B.
Sigma-Aldrich
Lignoceric acid, ≥99% (GC)