- Resolving the motional modes that code for RNA adaptation.
Resolving the motional modes that code for RNA adaptation.
Science (New York, N.Y.) (2006-02-04)
Qi Zhang, Xiaoyan Sun, Eric D Watt, Hashim M Al-Hashimi
PMID16456078
ABSTRACT
Using a domain elongation strategy, we decoupled internal motions in RNA from overall rotational diffusion. This allowed us to site-specifically resolve a manifold of motional modes in two regulatory RNAs from HIV-1 with the use of nuclear magnetic resonance spin relaxation methods. Base and sugar librations vary on a picosecond time scale and occur within helical domains that move collectively at diffusion-limited nanosecond time scales. Pivot points are short, functionally important, and highly mobile internal loops. These spontaneous changes in RNA conformation correlate quantitatively with those that follow adaptive recognition of diverse targets. Thus, ligands may stabilize existing RNA conformations rather than inducing new ones.
MATERIALS