MilliporeSigma
  • Levofloxacin and ciprofloxacin decrease procainamide and N-acetylprocainamide renal clearances.
Levofloxacin and ciprofloxacin decrease procainamide and N-acetylprocainamide renal clearances.
Antimicrobial agents and chemotherapy (2005-03-29)
Larry A Bauer, Douglas J Black, Jennifer S Lill, Julie Garrison, Vidmantas A Raisys, Thomas M Hooton
ABSTRACT
Ten healthy adults participated in a randomized, crossover drug interaction study testing procainamide only, procainamide plus levofloxacin, and procainamide plus ciprofloxacin. During levofloxacin therapy, most procainamide and N-acetylprocainamide (NAPA) pharmacokinetic parameters, including decreased renal clearances and renal clearance/creatinine clearance ratios, changed (P < 0.05). During ciprofloxacin treatment, only procainamide and NAPA renal clearances decreased significantly.
Sigma-Aldrich
N-Acetylprocainamide, ≥99%
Sigma-Aldrich
N-Acetylprocainamide hydrochloride, ≥99% (HPLC), powder